Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Microcope Slides, Igneous Rocks
SDPSCI_ED

Catalog No. S20727 Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20727 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20727 Supplier 3B Scientific™ Supplier No. W13150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3B Scientific™ Microcope Slides are thin sections of igneous rocks

31 slides including: altered granite, andesite, basalt, basalt with olivin, basalt with phenocryst and white feldspat, picrit basalt, tholeiitic basalt, granodiorite, pillow lava, dacite, diorite, diorite quartzique, dolerite, doreite, gabbro, granite, two-micas granite, and more

Specifications

Includes 31 Slides
Width (Metric) 4.5 cm
Height (Metric) 5.5 cm
Quantity 1
Material Glass, Rock
Color White, Gray
Languages English
For Use With (Application) Microscope
Subject Earth Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.