Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Microcope Slides, Sedimentary Rocks
SDPSCI_ED

Catalog No. S20729 Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S20729 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20729 Supplier 3B Scientific™ Supplier No. W13152
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3B Scientific™ Microcope Slides are thin sections of sedimentary rocks

22 slides including arkose, chalk, coal, gypsum, limestone with alveolina, limestone with asphalt, fossilized limestone, limestone with crinoid stem, glauconitic limestone, limestone with globotruncana (maestrichtien), limestone with globigerinina (paleocene), limestone with ooids, and more

Specifications

Includes 22 Slides
Width (Metric) 4.5 cm
Height (Metric) 5.5 cm
Quantity 1
Material Glass, Rock
Color White, Green
Languages English
For Use With (Application) Microscope
Subject Earth Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.