Learn More
Analytical Sales & Services MicroCross for .025" Tubing Sleeves
Catalog No. 5000807077
Encompass
Micro Cross Assembly for .025" Microtight Sleeves .006" Thru-Hole Swept Volume 29nL Includes (4) 55172 and (4) 59416 Fittings
Qty/Pkg: EACH
Catalog No. 50-008-07077 Supplier Analytical Sales & Services Supplier No. 59777
Micro Cross Assembly for .025" Microtight Sleeves .006" Thru-Hole Swept Volume 29nL Includes (4) 55172 and (4) 59416 Fittings
Qty/Pkg: EACH