Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Microcrystal Growth Kit
GREENER_CHOICESCI_ED

Catalog No. S37739 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Student
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S37739 1 Student
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid and Chromium/ hexavalent compounds, which are known to the State of California to cancer and reproductive harm.
Catalog No. S37739 Supplier Innovating Science™ Supplier No. IS5009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid and Chromium/ hexavalent compounds, which are known to the State of California to cancer and reproductive harm.

Includes

1 x 60 mL each of; Ammonium Phosphate Monobasic Solution, Copper (II) Sulfate Solution, Potassium Chromate Solution, Potassium Ferricyanide Solution, Sodium Chloride Solution, Sodium Nitrate Solution

Students will use different solutions to create a range of fun and exciting microcrystalline forms to discover the wonders of crystalline growth and properties.

  • Fun and exciting exercise for students
  • Microscope or magnifier is used to study crystals

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To create a range of fun and exciting microcrystalline forms to discover the wonders of crystalline growth and properties.
Includes 1 x 60 mL each of: Ammonium Phosphate Monobasic Solution, Copper (II) Sulfate Solution, Potassium Chromate Solution, Potassium Ferricyanide Solution, Sodium Chloride Solution, Sodium Nitrate Solution
Quantity 1
Dimensions (L x W x H) 5.5 x 4.25 x 6.75 in.
Class Size 1 Student
Product Type Microcrystal Growth Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.