Learn More
Buehler MICROCUT 8" PSA 1200GRIT 100PK
Catalog No. NC9780849
Encompass
This item has a minimum qty of 2 per supplier requirements.
CARBIMET Discs - PSA, Quantity:100, Grit:1200 (P2500), 8 in. (203mm) PSA backed, for wet or dry use
Catalog No. NC9780849 Supplier Buehler Supplier No. 36081200
This item has a minimum qty of 2 per supplier requirements.
CARBIMET Discs - PSA, Quantity:100, Grit:1200 (P2500), 8 in. (203mm) PSA backed, for wet or dry use