Learn More
Enzo Life Sciences Microcystin-HtyR (25µg)
SDP
Supplier:  Enzo Life Sciences ALX350174C025
Encompass_Preferred
Isolated from Microcystis aeruginosa. Microcystin containing a homotyrosine (Hty) in position 2 and arginine (R) in position 4. Hepatotoxic. Alternative name: [Hty2]microcystin-YR. Purity: ≥95% (HPLC). Appearance: White to off-white powder or oily film adhered to inside of the vial. Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8774