Learn More
Enzo Life Sciences Microcystin-LA (100µg). CAS: 96180-79-9
SDP
Catalog No. 502008728
Encompass_Preferred
Analog of microcystin-LR (ALX-350-012) with Ala substituted in place of Arg. Inhibits protein phosphatase 2A (PP2A) and protein phosphatase 3 (PP3) more potently than protein phosphatase 1 (PP1). Ozonation of toxic water samples does lead to a complete loss of the microcystins and their toxicity. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8728 Supplier Enzo Life Sciences Supplier No. ALX350096C100
Analog of microcystin-LR (ALX-350-012) with Ala substituted in place of Arg. Inhibits protein phosphatase 2A (PP2A) and protein phosphatase 3 (PP3) more potently than protein phosphatase 1 (PP1). Ozonation of toxic water samples does lead to a complete loss of the microcystins and their toxicity. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol or methanol. Long Term Storage: -20°C.