Learn More
Enzo Life Sciences Microcystin-LR, (1mg), CAS Number: 101043-37-2
SDP
Catalog No. NC9424956
Encompass_Preferred
Inhibitor of PP1 and PP2A. Heptapeptide ester hepatotoxin. Tumor promoter. Equally potent and selective inhibitor of protein phosphatase 1 (PP1) and 2A (PP2A). PP2B is less sensitive and PP2C is not inhibited up to 4µM. Useful for affinity-purification of PP2A. The product is not cell permeable except in liver cells, which appear to have a functional uptake system. Is absorbed by hepatocytes.7.7. Long Term Storage: -20°C
Catalog No. NC9424956 Supplier Enzo Life Sciences Supplier No. ALX350012M001
Inhibitor of PP1 and PP2A. Heptapeptide ester hepatotoxin. Tumor promoter. Equally potent and selective inhibitor of protein phosphatase 1 (PP1) and 2A (PP2A). PP2B is less sensitive and PP2C is not inhibited up to 4µM. Useful for affinity-purification of PP2A. The product is not cell permeable except in liver cells, which appear to have a functional uptake system. Is absorbed by hepatocytes.7.7. Long Term Storage: -20°C