Learn More
Enzo Life Sciences Microcystin-LW (25µg). CAS: 157622-02-1
SDP
Catalog No. 502008720
Encompass_Preferred
Analog of microcystin-LR (ALX-350-012) with Trp substituted in place of Arg. Hydrophobic and believed to be more cell permeable than other microcystins. May prove useful in biochemical studies in intact cells. Microcystin-LW has a characteristically different absorption spectrum compared to other microcystins, making it a useful reference compound for HPLC analysis. The Trp confers an absorption maximum at 222nm, whereas most microcystins have a characteristic maximum at 239nm. Purity: ≥95% (HPLC). Solubility: Soluble in 100% methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8720 Supplier Enzo Life Sciences Supplier No. ALX350080C025
Analog of microcystin-LR (ALX-350-012) with Trp substituted in place of Arg. Hydrophobic and believed to be more cell permeable than other microcystins. May prove useful in biochemical studies in intact cells. Microcystin-LW has a characteristically different absorption spectrum compared to other microcystins, making it a useful reference compound for HPLC analysis. The Trp confers an absorption maximum at 222nm, whereas most microcystins have a characteristic maximum at 239nm. Purity: ≥95% (HPLC). Solubility: Soluble in 100% methanol. Long Term Storage: -20°C.