Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microcystin-LW Solution , MilliporeSigma™ Supelco™

Catalog No. 111018004 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8004 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8004 Supplier MilliporeSigma Supelco Supplier No. 337631ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Microcystin-LW is a toxic analog of microcystin produced by cyanobacteria in freshwater systems.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD16661165
Synonym Biotoxin; Cyanobacterial toxin; Algae bloom toxin

Specifications

CAS 157622-02-1
Flash Point 11°C
Linear Formula C54H72N8O12
Quantity 1 mL
Formula Weight 1025.20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.