Learn More
Enzo Life Sciences Microcystin-LY (100µg). CAS: 123304-10-9
SDP
Supplier:  Enzo Life Sciences ALX350148C100
Encompass_Preferred
Analog of microcystin-LR (ALX-350-012) with Tyr substituted in place of Arg. Its toxicity is intermediate between the more toxic microcystin-LF and the less toxic microcystin-LR. Purity: ≥95% (HPLC). Appearance: Whitish film adhered to inside of the vial. Solubility: Soluble in 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8758