Learn More
Enzo Life Sciences Microcystin-RR (50µg). CAS: 111755-37-4
SDP
Catalog No. 502008702
Encompass_Preferred
Arg-Arg analog of microcystin-LR (ALX-350-012). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A). Purity: ≥95% (HPLC). Appearance: Clear to whitish film adhered to inside of vial. Solubility: Soluble in 80% aqueous methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8702 Supplier Enzo Life Sciences Supplier No. ALX350043C050
Arg-Arg analog of microcystin-LR (ALX-350-012). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A). Purity: ≥95% (HPLC). Appearance: Clear to whitish film adhered to inside of vial. Solubility: Soluble in 80% aqueous methanol. Long Term Storage: -20°C.