Learn More
Enzo Life Sciences Microcystin-RR (Analytical Standard) (10µg). CAS: 111755-37-4
SDP
Catalog No. 502008885
Encompass_Preferred
Microcystin-RR is the Arg-Arg analog of microcystin-LR (ALX-350-431). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A). This product can be used as an analytical standard for water testing or for molecular studies when high purity is required. Purity: ≥99% (HPLC). Appearance: Clear to whitish film adhered to inside of vial. Long Term Storage: -20°C.
Catalog No. 50-200-8885 Supplier Enzo Life Sciences Supplier No. ALX350432C010
Microcystin-RR is the Arg-Arg analog of microcystin-LR (ALX-350-431). Hepatotoxic, although found to be up to 10-fold less toxic than microcystin-LR on i.p. injection in mice. Potent inhibitor of protein phosphatase 2A (PP2A). This product can be used as an analytical standard for water testing or for molecular studies when high purity is required. Purity: ≥99% (HPLC). Appearance: Clear to whitish film adhered to inside of vial. Long Term Storage: -20°C.