Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microcystin-RR Solution, 10 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111018076 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8076 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8076 Supplier MilliporeSigma Supelco Supplier No. 335771ML
Add to Cart
Add to Cart

Microcystin-RR is a microcystin generally produced by cyanobacteria.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111755-37-4
Linear Formula C49H75N13O12
MDL Number MFCD00871765
Type Analytical Standard
Quantity 1 mL
Formula Weight 1038.20
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.