Learn More
Enzo Life Sciences Microcystin-WR (100µg). CAS: 138234-58-9
SDP
Supplier:  Enzo Life Sciences ALX350167C100
Encompass_Preferred
Purity: ≥95% (HPLC). Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8768