Learn More
ACL Inc Staticide 9" X 9", 150 Micrfodenier wipers per bag
Catalog No. NC1841773
Encompass_Preferred
95% Polester / 5% nylon Lint Free Super Soft, ultrasonically sealed edges for fiber & particle retention, free of silicone, amides and DOP contamination. ISO Class 4 -5 (Calss 10 - 100)
Catalog No. NC1841773 Supplier ACL Inc Supplier No. 8299MD
95% Polester / 5% nylon Lint Free Super Soft, ultrasonically sealed edges for fiber & particle retention, free of silicone, amides and DOP contamination. ISO Class 4 -5 (Calss 10 - 100)