Learn More
Instech Low-torque dual channel quartz-lined Microdialysis Swivel for Mice and Rats
SDP
Catalog No. 5080975
Encompass
The Instech 375/D/22QM is a low-torque, two-channel quartz-lined swivel designed for microdialysis in freely-moving mice or rats. This VAB™ compatible swivel features quartz lining on both the center and side channels to ensure the highest level of compound inertness and recovery. The stainless steel body provides durability for long-term use, while the low-torque design prevents tangling in delicate mouse tethers. Integrated universal swivel-to-tether clamps ensure a secure connection to your protective tether system.
Features
  • Application: Dual-channel microdialysis for mice and rats
  • Compatibility: Mates with multi-channel VAB™ tethers
  • Material: Quartz-lined center and side channels for inertness
  • Design: Low-torque mechanism optimized for mouse research
  • Durability: Stainless steel construction for a long service life
  • Access: Includes universal swivel-to-tether clamp
Catalog No. 50-809-75 Supplier Instech Supplier No. 375/D/22QM
The Instech 375/D/22QM is a low-torque, two-channel quartz-lined swivel designed for microdialysis in freely-moving mice or rats. This VAB™ compatible swivel features quartz lining on both the center and side channels to ensure the highest level of compound inertness and recovery. The stainless steel body provides durability for long-term use, while the low-torque design prevents tangling in delicate mouse tethers. Integrated universal swivel-to-tether clamps ensure a secure connection to your protective tether system.
Features
  • Application: Dual-channel microdialysis for mice and rats
  • Compatibility: Mates with multi-channel VAB™ tethers
  • Material: Quartz-lined center and side channels for inertness
  • Design: Low-torque mechanism optimized for mouse research
  • Durability: Stainless steel construction for a long service life
  • Access: Includes universal swivel-to-tether clamp