Learn More
Instech Microdialysis Swivel for Rats, quartz-lined center channel
SDP
Catalog No. 5080974
Encompass
Two Channel Stainless Steel Swivels
For Microdialysis with Rats
375/D/22QE - Two channel swivel with quartz-lined center channel for microdialysis with freely-moving rats. (Previously distributed by Harvard Apparatus / CMA as part no. 61-0004 / 830 9048.).
Its dual channels allow simultaneous infusion and withdrawal, independent dual‑line infusion, or infusion on one channel while monitoring on the other. Like the stainless single channel swivels, these swivels have been built to last a lifetime (however, users should follow the instructions carefully when cleaning the swivels after every use).
Each swivel includes a universal clamp compatible with button, harness, and head block tethers. Repair and replacement parts are available.
Features
  • Dual channel
  • Stainless steel body
  • Self-adjusting seals
  • Includes universal swivel-to-tether clamp
Catalog No. 50-809-74 Supplier Instech Supplier No. 375/D/22QE
Two Channel Stainless Steel Swivels
For Microdialysis with Rats
375/D/22QE - Two channel swivel with quartz-lined center channel for microdialysis with freely-moving rats. (Previously distributed by Harvard Apparatus / CMA as part no. 61-0004 / 830 9048.).
Its dual channels allow simultaneous infusion and withdrawal, independent dual‑line infusion, or infusion on one channel while monitoring on the other. Like the stainless single channel swivels, these swivels have been built to last a lifetime (however, users should follow the instructions carefully when cleaning the swivels after every use).
Each swivel includes a universal clamp compatible with button, harness, and head block tethers. Repair and replacement parts are available.
Features
  • Dual channel
  • Stainless steel body
  • Self-adjusting seals
  • Includes universal swivel-to-tether clamp