Learn More
Dr Instruments Microdissection Scissors - 9M 4.75", Stainless Steel.
SDP
Catalog No. NC1598781
Encompass_Preferred
Extremely fine and straight or curved blades for delicate dissection. Serrated handle with spring band for control. Overall length 4.75" with a 1/2" blade. Made of high quality stainless steel. Straigny blades.
This item is not returnable. View return policy
Catalog No. NC1598781 Supplier Dr Instruments Supplier No. 9M
Extremely fine and straight or curved blades for delicate dissection. Serrated handle with spring band for control. Overall length 4.75" with a 1/2" blade. Made of high quality stainless steel. Straigny blades.