Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Microdissection Forceps Set of 5, Super Fine Forceps, Stainless Steel
SDPSCI_ED

Catalog No. s72112
Change view
Click to view available options
Tip Style:
2 Curved, 3 Straight
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
S72112 2 Curved, 3 Straight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

California Proposition 65 WARNING: Cancer and Reproductive Harm

Catalog No. S72112 Supplier DR Instruments Supplier No. MDFKIT
Only null left
Add to Cart
Edge
Add to Cart

California Proposition 65 WARNING: Cancer and Reproductive Harm

Microdissection Forceps Set of 5 is ideal for super-fine dissection applications and working under a microscope.

  • Contains widely used 5 pairs of forceps
  • Highly precise and extra fine tips to perform micro-dissection
  • Includes 2 curved and 3 straight micro-dissection forceps
  • Stainless Steel
  • Transparent case

Specifications

Product Type Microdissection Forceps
Material Stainless Steel
For Use With (Application) Super-fine dissection
Quantity 5 Pk.
Tip Style 2 Curved, 3 Straight
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.