Learn More
RWD LIFE SCIENCE INC MICRODRILL HOLDER
SDP
Catalog No. NC2319381
Encompass
Holder secures the microdrill to sterotaxic instruments. Controls the depth of drilling by the manipulator. Easy and accurate operation protects brain tissue from damage caused by excessive drilling.Holds the microdrill with diameter about 14.5mm.
Catalog No. NC2319381 Supplier RWD LIFE SCIENCE INC Supplier No. 68605
Holder secures the microdrill to sterotaxic instruments. Controls the depth of drilling by the manipulator. Easy and accurate operation protects brain tissue from damage caused by excessive drilling.Holds the microdrill with diameter about 14.5mm.