Learn More
Sutter Instruments Microelectrode Beveler
SDP
Catalog No. NC1724231
Encompass_Preferred
Elegant and simple to use, the BV-10 offers precision beveling of micropipette tips between 0.1 and 100 µm. The unique abrasive plate drive system is vibration free for greater control of the beveling process. Beveling can be accomplished very rapidly and produces consistent tip diameters using the techniques as described by Brown and Flaming, Science, August 1974, Vol. 185.
Catalog No. NC1724231 Supplier Sutter Instruments Supplier No. FGBV10D
Add to Cart
Add to Cart
Request a Quote
Elegant and simple to use, the BV-10 offers precision beveling of micropipette tips between 0.1 and 100 µm. The unique abrasive plate drive system is vibration free for greater control of the beveling process. Beveling can be accomplished very rapidly and produces consistent tip diameters using the techniques as described by Brown and Flaming, Science, August 1974, Vol. 185.