Learn More
World Precision Instrument Microelectrode Holder (MEH2RW)-MEH2RW10
SDP
Catalog No. 502521135
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1135 Supplier World Precision Instrument Supplier No. MEH2RW10
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.