Learn More
World Precision Instrument Microelectrode Holder (MEH2RW)-MEH2RW12
SDP
Catalog No. 502521134
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1134 Supplier World Precision Instrument Supplier No. MEH2RW12
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.