Learn More
World Precision Instrument Microelectrode Holder (MEH2S)-MEH2S15
SDP
Catalog No. 502521130
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1130 Supplier World Precision Instrument Supplier No. MEH2S15
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.