Learn More
World Precision Instrument Microelectrode Holder (MEH2S)-MEH2S12
SDP
Catalog No. 502521131
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1131 Supplier World Precision Instrument Supplier No. MEH2S12
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.