Learn More
World Precision Instrument Microelectrode Holder (MEH2SW)-MEH2SW20
SDP
Catalog No. 502521125
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1125 Supplier World Precision Instrument Supplier No. MEH2SW20
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.