Learn More
World Precision Instrument Microelectrode Holder (MEH345)-MEH34512
SDP
Catalog No. 502521123
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1123 Supplier World Precision Instrument Supplier No. MEH34512
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.