Learn More
World Precision Instrument Microelectrode Holder (MEH3FW45)-MEH3FW4515
SDP
Catalog No. 502521120
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1120 Supplier World Precision Instrument Supplier No. MEH3FW4515
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.