Learn More
World Precision Instrument Microelectrode Holder (MEH3R)-MEH3R20
SDP
Catalog No. 502538716
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8716 Supplier World Precision Instrument Supplier No. MEH3R20
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.