Learn More
World Precision Instrument Microelectrode Holder (MEH3R)-MEH3R15
SDP
Catalog No. 502538717
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8717 Supplier World Precision Instrument Supplier No. MEH3R15
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.