Learn More
World Precision Instrument Microelectrode Holder (MEH3RF)-MEH3RF15
SDP
Catalog No. 502538714
Encompass
For use with these WPI ProductsSYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8714 Supplier World Precision Instrument Supplier No. MEH3RF15
For use with these WPI ProductsSYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.