Learn More
World Precision Instrument Microelectrode Holder (MEH3RF)-MEH3RF10
SDP
Catalog No. 502538715
Encompass
For use with these WPI ProductsSYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8715 Supplier World Precision Instrument Supplier No. MEH3RF10
For use with these WPI ProductsSYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.