Learn More
World Precision Instrument Microelectrode Holder (MEH3RFW) -MEH3RFW15
SDP
Catalog No. 502538712
Encompass
For use with these WPI Products:SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8712 Supplier World Precision Instrument Supplier No. MEH3RFW15
For use with these WPI Products:SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.