Learn More
World Precision Instrument Microelectrode Holder (MEH3RFW) -MEH3RFW12
SDP
Catalog No. 502521119
Encompass
For use with these WPI Products:SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1119 Supplier World Precision Instrument Supplier No. MEH3RFW12
For use with these WPI Products:SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.