Learn More
World Precision Instrument Microelectrode Holder (MEH3RW)-MEH3RW15
SDP
Catalog No. 502521117
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1117 Supplier World Precision Instrument Supplier No. MEH3RW15
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.