Learn More
World Precision Instrument Microelectrode Holder (MEH3S)-MEH3S12
SDP
Catalog No. 502521115
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1115 Supplier World Precision Instrument Supplier No. MEH3S12
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.