Learn More
World Precision Instrument Microelectrode Holder (MEH3S)-MEH3S20
SDP
Catalog No. 502521113
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1113 Supplier World Precision Instrument Supplier No. MEH3S20
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.