Learn More
World Precision Instrument Microelectrode Holder (MEH3SBW)-MEH3SBW10
SDP
Catalog No. 502538711
Encompass
For use with these WPI Products: ISO-80, ISO-DAM8A
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8711 Supplier World Precision Instrument Supplier No. MEH3SBW10
For use with these WPI Products: ISO-80, ISO-DAM8A
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.