Learn More
World Precision Instrument Microelectrode Holder (MEH3SBW)-MEH3SBW15
SDP
Catalog No. 502538710
Encompass
For use with these WPI Products: ISO-80, ISO-DAM8A
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8710 Supplier World Precision Instrument Supplier No. MEH3SBW15
For use with these WPI Products: ISO-80, ISO-DAM8A
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.