Learn More
World Precision Instrument Microelectrode Holder (MEH3SFW)-MEH3SFW20
SDP
Catalog No. 502521105
Encompass
For use with these WPI Products: SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1105 Supplier World Precision Instrument Supplier No. MEH3SFW20
For use with these WPI Products: SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.