Learn More
World Precision Instrument Microelectrode Holder (MEH3SW)-MEH3SW10
SDP
Catalog No. 502521104
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1104 Supplier World Precision Instrument Supplier No. MEH3SW10
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.