Learn More
World Precision Instrument Microelectrode Holder (MEH3W45)-MEH3W4520
SDP
Catalog No. 502521099
Encompass
For use with these WPI Products: SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1099 Supplier World Precision Instrument Supplier No. MEH3W4520
For use with these WPI Products: SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.