Learn More
World Precision Instrument Microelectrode Holder (MEH6SF)-MEH6SF15
SDP
Supplier:  World Precision Instrument MEH6SF15
Encompass
For use with these WPI Products: SYS-705, SYS-773, 767, 721, FD223
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8706