Learn More
World Precision Instrument Microelectrode Holder (MEH7)-MEH715
SDP
Catalog No. 502538700
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8700 Supplier World Precision Instrument Supplier No. MEH715
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.