Learn More
World Precision Instrument Microelectrode Holder (MEH7)-MEH712
SDP
Catalog No. 502538701
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8701 Supplier World Precision Instrument Supplier No. MEH712
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.