Learn More
World Precision Instrument Microelectrode Holder (MEH7)-MEH710
SDP
Catalog No. 502538702
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8702 Supplier World Precision Instrument Supplier No. MEH710
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.