Learn More
World Precision Instrument Microelectrode Holder (MEH7)-MEH720
SDP
Catalog No. 502538699
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8699 Supplier World Precision Instrument Supplier No. MEH720
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.