Learn More
World Precision Instrument Microelectrode Holder (MEH7W)-MEH7W12
SDP
Catalog No. 502538697
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8697 Supplier World Precision Instrument Supplier No. MEH7W12
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.