Learn More
World Precision Instrument Microelectrode Holder (MEH7W)-MEH7W10
SDP
Catalog No. 502538698
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-253-8698 Supplier World Precision Instrument Supplier No. MEH7W10
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.